Informational:Informational:This product is animal origin and can only be shipped within the United States
Thermo Scientific Chemicals

Trypsin Inhibitor, chicken egg whites

Catalog number: J63927.03
1 g, Each
Thermo Scientific Chemicals

Trypsin Inhibitor, chicken egg whites

Catalog number: J63927.03
1 g, Each
Quantity
Catalog number: J63927.03
also known as J63927-03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
9035-81-8
Form
Powder
Assay (special)
1 mg inhibits ≥1.20 mg trypsin
Assay (special)
Protein: Report assay value

Description

Soybean trypsin inhibior (STI) is a protein which reduces trypsin (a proteolytic enzyme) as well as plasma kallikrein, factor Xa and plasmin activity. Soybean trypsin inhibitor has demonstrated inhibition of human neutrophil elastase at an IC50=0.3 nM. At this concentration soybean trypsin inhibitor showed neither cytotoxic nor haemolytic effects on populations of human blood cells. In fibrin matrix, and without toxic effects, STI had a strong inhibitory effect on human umbilical vein endothelial cells migration and tubulogenesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text