Thermo Scientific Chemicals

Citrate, 0.5M buffer soln., pH 6.0

Catalog number: J63950.AP
500 mL, Each
Thermo Scientific Chemicals

Citrate, 0.5M buffer soln., pH 6.0

Catalog number: J63950.AP
500 mL, Each
Quantity
Catalog number: J63950.AP
also known as J63950-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6132-04-3
IUPAC Name
trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
Molecular Formula
C6H9Na3O9
InChI Key
NLJMYIDDQXHKNR-UHFFFAOYSA-K
SMILES
O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Form
Clear Liquid
Comment
Filtered through 0.22 micron filter
pH
6.0 ± 0.2

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text