Thermo Scientific Chemicals

Ethyl Violet, Thermo Scientific Chemicals

Catalog number: J63951.14
25 g, Each
Thermo Scientific Chemicals

Ethyl Violet, Thermo Scientific Chemicals

Catalog number: J63951.14
25 g, Each
Quantity
Catalog number: J63951.14
also known as J63951-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2390-59-2
IUPAC Name
tris[4-(diethylamino)phenyl]methylium chloride
Molecular Formula
C31H42ClN3
InChI Key
JVICFMRAVNKDOE-UHFFFAOYSA-M
SMILES
[Cl-].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1)N(CC)CC
Appearance (Color)
Dark brown to green
Loss on Drying
≤5.0%
Insoluble material
≤2.0%
Solution Test
Dark purple solution (1mg/mL, water)
Form
Powder

Description

Ethyl violet has been used to stain pancreatic tissue and elastin in the detection of gram negative bacteria, and as a reagent for the extractive-spectrophotometric determination of anionic surfactants. It has been used in the extraction and spectrophotometric determination of copper. Along with the anionic dye zincon, ethyl violet is used in counterion dye of proteins in SDS-polyacrylamide gel electrophoresis. Along with the anionic dye zincon, ethyl violet is used in counterion dye of proteins in SDS-polyacrylamide gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl violet has been used to stain pancreatic tissue and elastin in the detection of gram negative bacteria, and as a reagent for the extractive-spectrophotometric determination of anionic surfactants. It has been used in the extraction and spectrophotometric determination of copper. Along with the anionic dye zincon, ethyl violet is used in counterion dye of proteins in SDS-polyacrylamide gel electrophoresis. Along with the anionic dye zincon, ethyl violet is used in counterion dye of proteins in SDS-polyacrylamide gel electrophoresis.

Solubility
Soluble in water (10 mg/ml).

Notes
Dye content approx. 80%
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text