Thermo Scientific Chemicals

Ticlopidine Hydrochloride

Catalog number: J63971.06
5 g, Each
Thermo Scientific Chemicals

Ticlopidine Hydrochloride

Catalog number: J63971.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J63971.06
also known as J63971-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
53885-35-1
IUPAC Name
hydrogen 5-[(2-chlorophenyl)methyl]-4H,5H,6H,7H-thieno[3,2-c]pyridine chloride
Molecular Formula
C14H15Cl2NS
InChI Key
MTKNGOHFNXIVOS-UHFFFAOYSA-N
SMILES
[H+].[Cl-].ClC1=CC=CC=C1CN1CCC2=C(C1)C=CS2
Assay (unspecified)
99-101%
Water Content (Karl Fischer Titration)
0.5% max.
Sulfated Ash
0.1% max.

Description

Ticlopidine hydrochloride is a selective P2Y12 receptor antagonist. It inhibits ADP-induced platelet aggregation and displays antithrombotic activity following oral administration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text