Thermo Scientific Chemicals

Dynorphin A (1-13), Porcine

Catalog number: J64015.MCR
1 mg, Each
Thermo Scientific Chemicals

Dynorphin A (1-13), Porcine

Catalog number: J64015.MCR
1 mg, Each
Quantity
Catalog number: J64015.MCR
also known as J64015-M^
Price (USD)
Quantity
-
Have Questions?
Assay from Suppliers CofA
Peptide Purity: Report Value
Solution Test
1 mg/ml in water
Form
Lyophilized powder
Mass Spec
Exhibits correct MW

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text