1x1 image pixel for data collection
Cellulase, chromatographically purified, T. reesei
Cellulase, chromatographically purified, T. reesei
Thermo Scientific Chemicals

Cellulase, chromatographically purified, T. reesei

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number J64019.03
also known as J64019-03
Price (USD)
190.65
Online Exclusive
212.00
Save 21.35 (10%)
Each
Estimated availability date 11-Jul-2025
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
190.65
Online Exclusive
212.00
Save 21.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS9012-54-8
SpecificationsSpecification SheetSpecification Sheet
Specific Absorbance (E 1%/1cm)A280nm (1 mg/ml) : Reported assay value
Assay from Supplier's CofA≥45 units per mg (dry weight)
CommentOne Unit liberates 0.01 mg of glucose per hour from microcrystalline cellulose at 37°C. pH 5.0
Enzymatic ActivityLipase : Reported assay value
Enzymatic ActivityNuclease : Reported assay value
View more
Ask our AI about this Product
Cellulase has been used to degrade cello-oligosaccharides into glucose and to study the biodegradability of bioabsorbable bacterial cellulose (BBC) material. It has been used in a study to assess the differential regulation of the cellulase transcription factors XYR1, ACE2, and ACE. It is also applied in the biodegradation of paper products such as foolscap paper, filter paper, newspaper and office paper as well as microcrystalline cellulose. It is added to detergents to improve cleansing properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
T01L058Certificate of AnalysisJul 02, 2025J64019.03, J64019.MD
R08L012Certificate of AnalysisMay 17, 2025J64019.03, J64019.MD
P05L033Certificate of AnalysisMar 10, 2025J64019.03, J64019.MD
X18K028Certificate of AnalysisNov 05, 2024J64019.03, J64019.MD
W11K022Certificate of AnalysisSep 23, 2024J64019.03, J64019.MD
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection