Azido-PEG4-Acid is amine- or phosphine-reactive building block used to derivative primary amines for ligation to alkyne-tagged molecules.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Azido-PEG4-Acid is amine- or phosphine-reactive building block used to derivative primary amines for ligation to alkyne-tagged molecules.
Solubility
Soluble in water.
Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only
Hilde van Hattum.; Kim van der Zwaluw.; Gerben M. Visser.; Jos van Putten.; Rob Ruijtenbeek.; Roland J. Pieters. Functional assay for shiga-like toxin via detection by antibody capture and multivalent galabiose binding. Bioorganic & Medicinal Chemistry Letters. 2012, 22, (24), 7448-7450.