Thermo Scientific Chemicals

2'-Fluoro-2'-deoxyguanosine, 99%

Catalog number: J64055.03
1 g, Each
Thermo Scientific Chemicals

2'-Fluoro-2'-deoxyguanosine, 99%

Catalog number: J64055.03
1 g, Each
Quantity
Catalog number: J64055.03
also known as J64055-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78842-13-4
IUPAC Name
2-amino-9-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
Molecular Formula
C10H12FN5O4
InChI Key
UXUZARPLRQRNNX-SJLGBTOWNA-N
SMILES
NC1=NC2=C(N=CN2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2F)C(=O)N1
Water Content
≤8.0%
Appearance (Color)
White to off-white
Proton NMR
Conforms
Form
Powder
Purity
≥98.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text