Chemical Identifiers
CAS78842-13-4
IUPAC Name2-amino-9-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
Molecular FormulaC10H12FN5O4
InChI KeyUXUZARPLRQRNNX-SJLGBTOWNA-N
SMILESNC1=NC2=C(N=CN2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2F)C(=O)N1
View more