Thermo Scientific Chemicals

N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-fluoro-2'-deoxyadenosine-3'-(2-cyanoethyl diisopropylphosphoramidite), 98%

Catalog number: J64064.03
1 g, Each
Thermo Scientific Chemicals

N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-fluoro-2'-deoxyadenosine-3'-(2-cyanoethyl diisopropylphosphoramidite), 98%

Catalog number: J64064.03
1 g, Each
Quantity
Catalog number: J64064.03
also known as J64064-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
136834-22-5
IUPAC Name
N-{9-[(2R,3R,4R,5R)-4-[bis(4-methoxyphenyl)(phenyl)methoxy]-5-[({[bis(propan-2-yl)amino](2-cyanoethoxy)phosphanyl}oxy)methyl]-3-fluorooxolan-2-yl]-9H-purin-6-yl}benzamide
Molecular Formula
C47H51FN7O7P
InChI Key
NZDMBUCNULHYQM-JRJFZAHINA-N
SMILES
COC1=CC=C(C=C1)C(O[C@@H]1[C@@H](COP(OCCC#N)N(C(C)C)C(C)C)O[C@H]([C@@H]1F)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12)(C1=CC=CC=C1)C1=CC=C(OC)C=C1
Appearance (Color)
White to off-white
Assay from Supplier's CofA
≥97.5%
Form
Powder or crystals or crystalline powder or flakes
Proton NMR
Conforms to structure

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text