Thermo Scientific Chemicals

(S)-(+)-2-Amino-4-phosphonobutyric acid, 97%, Thermo Scientific Chemicals

Catalog number: J64075.MA
10 mg, Each
Thermo Scientific Chemicals

(S)-(+)-2-Amino-4-phosphonobutyric acid, 97%, Thermo Scientific Chemicals

Catalog number: J64075.MA
10 mg, Each
Quantity
Catalog number: J64075.MA
also known as J64075-MA
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
23052-81-5
IUPAC Name
(2S)-2-azaniumyl-4-phosphonatobutanoate
Molecular Formula
C4H8NO5P
InChI Key
DDOQBQRIEWHWBT-VKHMYHEASA-L
SMILES
[NH3+][C@@H](CCP([O-])([O-])=O)C([O-])=O
Optical Rotation
Report value
Form
Powder
Purity
≥96.0%

Description

A NMDA agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text