Cathepsin K Substrate (fluorogenic)
Cathepsin K Substrate (fluorogenic)
Thermo Scientific Chemicals
Cathepsin K Substrate (fluorogenic)
For cellular and molecular biology applications
Have Questions?
Change viewbuttonViewtableView
Substrate Quantity:
25mg
10mg
Catalog number J64082.MB
also known as J64082-MB
Price (USD)/ Each
317.00
-
Add to cart
Substrate Quantity:
25mg
Request bulk or custom format
Price (USD)/ Each
317.00
Add to cart
Cathepsin K Substrate (fluorogenic)
Catalog numberJ64082.MB
Price (USD)/ Each
317.00
-
Add to cart
Specifications
Recommended StorageStore at -20°C
ColorWhite
FormSolid
For Use With (Application)A sensitive fluorogenic substrate for cathepsin K
Substrate TypeCathepsin Substrate
View more
A sensitive fluorogenic substrate for cathepsin K