N-Acetyl-L-tryptophan, 99%, Thermo Scientific Chemicals
N-Acetyl-L-tryptophan, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Acetyl-L-tryptophan, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number J64096.14
also known as J64096-14
Price (USD)/ Each
98.30
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
98.30
Add to cart
N-Acetyl-L-tryptophan, 99%, Thermo Scientific Chemicals
Catalog numberJ64096.14
Price (USD)/ Each
98.30
-
Add to cart
Chemical Identifiers
CAS1218-34-4
IUPAC Name(2S)-2-acetamido-3-(1H-indol-3-yl)propanoic acid
Molecular FormulaC13H14N2O3
InChI KeyDZTHIGRZJZPRDV-LBPRGKRZSA-N
SMILESCC(=O)N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥98.5%
Optical Rotation+26 ± 2° (C=1, 1N NaOH)
FormWhite to off-white powder
Melting Point178.0-206.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only