GAP 27
Thermo Scientific Chemicals

GAP 27

For cellular and molecular biology applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 mg
1 mg
Catalog number J64109.EXD
also known as J64109-!D
Price (USD)/ Each
74.20
-
Add to cart
Quantity:
0.5 mg
Request bulk or custom format
Price (USD)/ Each
74.20
Add to cart
GAP 27
Catalog numberJ64109.EXD
Price (USD)/ Each
74.20
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Purification MethodHPLC
FormatSolid
Molecular Weight (g/mol)1305.54
Product TypeGAP 27
Quantity0.5 mg
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C