Thermo Scientific Chemicals

Chlorpropamide, Thermo Scientific Chemicals

Catalog number: J64110.22
100 g, Each
Thermo Scientific Chemicals

Chlorpropamide, Thermo Scientific Chemicals

Catalog number: J64110.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J64110.22
also known as J64110-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
94-20-2
IUPAC Name
1-(4-chlorobenzenesulfonyl)-3-propylurea
Molecular Formula
C10H13ClN2O3S
InChI Key
RKWGIWYCVPQPMF-UHFFFAOYSA-N
SMILES
CCCNC(=O)NS(=O)(=O)C1=CC=C(Cl)C=C1
Melting Point
126-130?C
Assay (unspecified)
98.0% min.

Description

Chlorpropamide is used in the treatment of diabetes metilus type 2. It acts to increase the secretion of insulin and is not effective in patients who do not have pancreatic beta cell function.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorpropamide is used in the treatment of diabetes metilus type 2. It acts to increase the secretion of insulin and is not effective in patients who do not have pancreatic beta cell function.

Solubility
Soluble in ethanol (1:12), acetone (1:5), chloroform (1:9) and solutions of alkali hydroxides. Does not mix well with water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text