Lck Inhibitor II
Thermo Scientific Chemicals

Lck Inhibitor II

 
Have Questions?
Quantity:
5 mg
Catalog number J64111.LB0
also known as J64111-#0
Price (USD)/ Each
319.00
-
Add to cart
Quantity:
5 mg
Request bulk or custom format
Price (USD)/ Each
319.00
Add to cart
Lck Inhibitor II
Catalog numberJ64111.LB0
Price (USD)/ Each
319.00
-
Add to cart
Chemical Identifiers
CAS918870-43-6
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Appearance (Color)White to off-white
Purity≥95.0%
A cell-permeable, ATP binding site-targeting, tri-substituted pyrimidine that acts as a highly potent Lck (lymphocyte specific kinase) inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
2°C to 8°C