Azadibenzocyclooctyne-PEG4-NHS ester, Thermo Scientific Chemicals
Azadibenzocyclooctyne-PEG4-NHS ester, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Azadibenzocyclooctyne-PEG4-NHS ester, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
10 mg
100 mg
Catalog number J64126.MB
also known as J64126-MB
Price (USD)/ Each
293.00
-
Add to cart
Quantity:
25 mg
Request bulk or custom format
Price (USD)/ Each
293.00
Add to cart
Azadibenzocyclooctyne-PEG4-NHS ester, Thermo Scientific Chemicals
Catalog numberJ64126.MB
Price (USD)/ Each
293.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialAzadibenzocyclooctyne-PEG4-NHS ester
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Storage Note 1-30° to -10°C
View more
Azadibenzocyclooctyne-PEG4-NHS ester is applied for protein-peptide conjugation, peptide-small molecule conjugation, 18F radiolabeling, protein-oligonucleotide conjugation and surface modification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azadibenzocyclooctyne-PEG4-NHS ester is applied for protein-peptide conjugation, peptide-small molecule conjugation, 18F radiolabeling, protein-oligonucleotide conjugation and surface modification.

Solubility
Soluble in DCM, DMF, DMSO and THF.

Notes
Moisture sensitive. Incompatible with oxidizing agents and heat.
RUO – Research Use Only

General References:

  1. Dingbin Liu; Jie Yang; He-Fang Wang; Zhongliang Wang; Xinglu Huang; Zhantong Wang; Gang Niu; A.R.Hight Walker; Xiaoyuan Chen. Glucose oxidase-catalyzed growth of gold nanoparticles enables quantitative detection of attomolar cancer biomarkers. Anal. Chem. 2014, 86,(12), 5800-5806.
  2. Guoquan Li; Yan Xing; Jing Wang; Peter S.Conti; Kai Chen. Near-infrared fluorescence imaging of CD13 receptor expression using a novel Cy5.5-labeled dimeric NGR peptide. Amino Acids. 2014, 46,(6), 1547-1556.