Cytidine-5'-diphosphocholine sodium salt
Cytidine-5'-diphosphocholine sodium salt
Thermo Scientific Chemicals

Cytidine-5'-diphosphocholine sodium salt

Intermediate in the synthesis of phosphatidylcholine; also a phospholipase inhibitor
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
50 g
Catalog number J64161.14
also known as J64161-14
Price (USD)/ Each
141.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
141.00
Add to cart
Cytidine-5'-diphosphocholine sodium salt
Catalog numberJ64161.14
Price (USD)/ Each
141.00
-
Add to cart
Chemical Identifiers
CAS33818-15-4
IUPAC Namesodium 2-(trimethylazaniumyl)ethyl ({[(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Molecular FormulaC14H25N4NaO11P2
InChI KeyYWAFNFGRBBBSPD-OCMLZEEQSA-M
SMILES[Na+].C[N+](C)(C)CCOP([O-])(=O)OP([O-])(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=CC(N)=NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)95.0% min.
Assay (unspecified)Solubility: 5% w/v in water
Loss on Drying6.0% max.
Cytidine-5'-diphosphocholine sodium salt hydrate acts as a neuroprotective agent in situations of hypoxia and ischemia. It is used as biosynthetic intermediate of membrane phospholipids. It is a cerebral vasodilator and inhibits PLA2 activation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cytidine-5′-diphosphocholine sodium salt hydrate acts as a neuroprotective agent in situations of hypoxia and ischemia. It is used as biosynthetic intermediate of membrane phospholipids. It is a cerebral vasodilator and inhibits PLA2 activation.

Solubility
Soluble in water (100 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. W.M.Clark; S.J.Warach; L.C.Pettigrew; R.E.Gammans; L.A.Sabounjian. A randomized dose-response trial of citicoline in acute ischemic stroke patients. Citicoline Stroke Study Group. Neurology. 1997, 49,(3), 671-678.
  2. S.Warach, MD, PhD; L.Creed Pettigrew, MD; J.F.Dashe, MD, PhD; P.Pullicino MD; David M.Lefkowitz, MD; L.Sabounjian, RN; K.Harnett, MS; U.Schwiderski, PhD; R.Gammans, PhD. Effect of citicoline on ischemic lesions as measured by diffusion-weighted magnetic resonance imaging. Annals of Neurology. 2000, 48,(5), 713-722.