Ammonium sulfate, 99+%, Molecular Biology Grade
Ammonium sulfate, 99+%, Molecular Biology Grade
Thermo Scientific Chemicals

Ammonium sulfate, 99+%, Molecular Biology Grade

 
Have Questions?
Quantity:
1 kg
Catalog number J64180.A1
also known as J64180-A1
Price (USD)/ Each
104.00
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
104.00
Add to cart
Ammonium sulfate, 99+%, Molecular Biology Grade
Catalog numberJ64180.A1
Price (USD)/ Each
104.00
-
Add to cart
Chemical Identifiers
CAS7783-20-2
IUPAC Namesulfuric acid diamine
Molecular FormulaH8N2O4S
InChI KeyBFNBIHQBYMNNAN-UHFFFAOYSA-N
SMILESN.N.OS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride content≤0.001%
Assay from Supplier's CofA≥99.0 to ≤101.0%
DNaseNot detected
Loss on Drying≤0.5%
Sulfated Ash≤0.005%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only