Centrophenoxine hydrochloride, 98%, Thermo Scientific Chemicals
Centrophenoxine hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Centrophenoxine hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J64196.06
also known as J64196-06
Price (USD)/ Each
46.70
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
46.70
Add to cart
Centrophenoxine hydrochloride, 98%, Thermo Scientific Chemicals
Catalog numberJ64196.06
Price (USD)/ Each
46.70
-
Add to cart
Specifications
Chemical Name or MaterialCentrophenoxine hydrochloride
CAS3685-84-5
Health Hazard 1H302
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3P264b-P270-P301+P312-P330-P501c
View more
Ester of dimethylethanolamine and 4-chlorophenoxyacetic acid; activates acetylcholinesterase activity in hippocampus of aged rats

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ester of dimethylethanolamine and 4-chlorophenoxyacetic acid; activates acetylcholinesterase activity in hippocampus of aged rats
RUO – Research Use Only