Thermo Scientific Chemicals

Cefaclor, Thermo Scientific Chemicals

Catalog number: J64252.09
10 g, Each
Thermo Scientific Chemicals

Cefaclor, Thermo Scientific Chemicals

Catalog number: J64252.09
10 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J64252.09
also known as J64252-09
Price (USD)
Quantity
-
Have Questions?

Description

Cefaclor is used to study urinary tract, intra-abdominal, and Haemophilus influenzae infections. It is used to study the mechanism of human renal organic anion and peptide transporters such as hOAT1, hPEPT1, and hPEPT2 and to study the effects of inhibition of penicillin-binding proteins on bacterial cell wall mucopeptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cefaclor is used to study urinary tract, intra-abdominal, and Haemophilus influenzae infections. It is used to study the mechanism of human renal organic anion and peptide transporters such as hOAT1, hPEPT1, and hPEPT2 and to study the effects of inhibition of penicillin-binding proteins on bacterial cell wall mucopeptide synthesis.

Solubility
Soluble in 1N HCl

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text