2-Amino-2'-deoxyadenosine, 99%
2-Amino-2'-deoxyadenosine, 99%
Thermo Scientific Chemicals

2-Amino-2'-deoxyadenosine, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number J64266.14
also known as J64266-14
Price (USD)/ Each
128.65
Online exclusive
143.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
128.65
Online exclusive
143.00 
Save 14.35 (10%)
Add to cart
2-Amino-2'-deoxyadenosine, 99%
Catalog numberJ64266.14
Price (USD)/ Each
128.65
Online exclusive
143.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
CAS4546-70-7
IUPAC Name(2R,3S,5R)-5-(2,6-diamino-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Molecular FormulaC10H14N6O3
InChI KeyNOLHIMIFXOBLFF-DWFKHXNSNA-N
SMILESNC1=NC(N)=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Solution TestClear colorless solution (20 mg/mL in water)
Assay from Supplier's CofA≥98.5% (HPLC)
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only