Sucrose, ultrapure, 99%
Sucrose, ultrapure, 99%
Thermo Scientific Chemicals

Sucrose, ultrapure, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
Catalog number J64270.22
also known as J64270-22
Price (USD)/ Each
15.65
Online exclusive
17.20 
Save 1.55 (9%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
15.65
Online exclusive
17.20 
Save 1.55 (9%)
Add to cart
Sucrose, ultrapure, 99%
Catalog numberJ64270.22
Price (USD)/ Each
15.65
Online exclusive
17.20 
Save 1.55 (9%)
-
Add to cart
Chemical Identifiers
CAS57-50-1
IUPAC Name(2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC12H22O11
InChI KeyCZMRCDWAGMRECN-PWPRYFECNA-N
SMILESOC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay from Supplier's CofA≥98.5%
Impurity contentSulfate, SO4: ≤0.006% (≤60ppm)
Optical Rotation+66.3 to +66.8°
Heavy Metals (as Pb)≤0.0005% (≤5ppm)
View more
Sucrose is used to prepare density gradients for cell/organelle separation. In addition, sucrose can be used as a supplement in plant, insect, and bacterial culture media. It can also be used in various enzymatic assays. Sucrose is used in molecular biology for the preparation of density gradients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sucrose is used to prepare density gradients for cell/organelle separation. In addition, sucrose can be used as a supplement in plant, insect, and bacterial culture media. It can also be used in various enzymatic assays. Sucrose is used in molecular biology for the preparation of density gradients.

Solubility
Soluble in water(1970g/L).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only