Chemical Identifiers
CAS14999-52-1
IUPAC Namedisodium [(2R,3S,5R)-3-hydroxy-5-(6-oxo-6,9-dihydro-1H-purin-9-yl)oxolan-2-yl]methyl phosphate
Molecular FormulaC10H11N4Na2O7P
InChI KeyVDPLPYMIJCLJEF-DZVPOJRENA-L
SMILES[Na+].[Na+].O[C@H]1C[C@@H](O[C@@H]1COP([O-])([O-])=O)N1C=NC2=C1N=CNC2=O
View more