Thermo Scientific Chemicals

Syk Inhibitor

Catalog number: J64281.LB0
5 mg, Each
Thermo Scientific Chemicals

Syk Inhibitor

Catalog number: J64281.LB0
5 mg, Each
Quantity
Catalog number: J64281.LB0
also known as J64281-#0
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
622387-85-3
IUPAC Name
3-[(1-methyl-1H-indol-3-yl)methylidene]-2-oxo-2,3-dihydro-1H-indole-5-sulfonamide
Molecular Formula
C18H15N3O3S
InChI Key
MLKHXLFEYOOYEY-UHFFFAOYSA-N
SMILES
CN1C=C(C=C2C(=O)NC3=C2C=C(C=C3)S(N)(=O)=O)C2=CC=CC=C12
Purity
≥95.0% (sum of two isomers)
Form
Solid

Description

A cell-permeable oxindole compound that acts as a potent, reversible, and ATP-competitive Syk inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text