Thermo Scientific™

Caspase-1 Inhibitor I

Catalog number: J64407.MCR
1 mg, Each
Thermo Scientific™

Caspase-1 Inhibitor I

Catalog number: J64407.MCR
1 mg, Each
Quantity
Catalog number: J64407.MCR
also known as J64407-M^
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
143313-51-3
IUPAC Name
(3S)-3-{[(2S)-1-[(2S)-2-[(2S)-2-acetamido-3-(4-hydroxyphenyl)propanamido]-3-methylbutanamido]-1-oxopropan-2-yl]amino}-4-oxobutanoic acid
Molecular Formula
C23H32N4O8
InChI Key
KQCBPHUOVBKBPQ-SJVNDZIOSA-N
SMILES
CC(C)[C@H](NC(=O)[C@H](CC1=CC=C(O)C=C1)NC(C)=O)C(=O)NC(=O)[C@H](C)N[C@@H](CC(O)=O)C=O
Assay by NMR
Consistent with structure
Form
Powder or crystals or crystalline powder
Appearance (Color)
White to off-white
Assay from Suppliers CofA
≥95%

Description

A potent, specific, and reversible inhibitor of caspases-1, -4 and -5

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text