Thermo Scientific Chemicals

PARP Inhibitor VIII, PJ34

Catalog number: J64413.LB0
5 mg, Each
Thermo Scientific Chemicals

PARP Inhibitor VIII, PJ34

Catalog number: J64413.LB0
5 mg, Each
Quantity
Catalog number: J64413.LB0
also known as J64413-#0
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
344458-15-7
IUPAC Name
2-(dimethylamino)-N-(6-oxo-5,6-dihydrophenanthridin-2-yl)acetamide hydrochloride
Molecular Formula
C17H18ClN3O2
InChI Key
RURAZZMDMNRXMI-UHFFFAOYSA-N
SMILES
Cl.CN(C)CC(=O)NC1=CC=C2NC(=O)C3=CC=CC=C3C2=C1
Purity
≥97.5% (HPLC)
Form
Solid
Appearance (Color)
White to pale brown

Description

A cell-permeable, water-soluble phenanthridinone-derivative that acts as a potent inhibitor of poly(ADP-ribose) polymerase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A cell-permeable, water-soluble phenanthridinone-derivative that acts as a potent inhibitor of poly(ADP-ribose) polymerase
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text