Thermo Scientific Chemicals

Ammonium sulfate, ultrapure, 99+%, Thermo Scientific Chemicals

Catalog number: J64419.36
500 g, Each
Thermo Scientific Chemicals

Ammonium sulfate, ultrapure, 99+%, Thermo Scientific Chemicals

Catalog number: J64419.36
500 g, Each
Quantity
Catalog number: J64419.36
also known as J64419-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7783-20-2
IUPAC Name
sulfuric acid diamine
Molecular Formula
H8N2O4S
InChI Key
BFNBIHQBYMNNAN-UHFFFAOYSA-N
SMILES
N.N.OS(O)(=O)=O
Residue on ignition
≤0.005%
Impurity content
Phosphate: ≤5 ppm
Chloride content
≤5 ppm
Impurity content
Nitrate: ≤0.001%
pH
5.0-6.0

Description

Ammonium sulfate is a widely used reagent in molecular biology and chromatography. It is useful in the isolation and purification of enzymes and proteins. It is used for the precipitation or fractionation of proteins or for purification of antibodies. It is also useful for crystallographic analysis of nucleic acids and proteins. Ammonium sulfate is also widely used in HPLC of proteins, such as in hydrophobic interaction chromatography. Ammonium sulfate is used in long PCR buffer, in PCR lysis solution, and in secondstrand cDNA synthesis. Also it is used for the HPLC analysis of phospholipids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium sulfate is a widely used reagent in molecular biology and chromatography. It is useful in the isolation and purification of enzymes and proteins. It is used for the precipitation or fractionation of proteins or for purification of antibodies. It is also useful for crystallographic analysis of nucleic acids and proteins. Ammonium sulfate is also widely used in HPLC of proteins, such as in hydrophobic interaction chromatography. Ammonium sulfate is used in long PCR buffer, in PCR lysis solution, and in secondstrand cDNA synthesis. Also it is used for the HPLC analysis of phospholipids.

Solubility
Soluble freely in water, Insoluble in alcohol and acetone.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text