Locostatin, Thermo Scientific Chemicals
Locostatin, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Locostatin, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
10 mg
Catalog number J64425.MF
also known as J64425-MF
Price (USD)/ Each
448.00
-
Add to cart
Quantity:
50 mg
Request bulk or custom format
Price (USD)/ Each
448.00
Add to cart
Locostatin, Thermo Scientific Chemicals
Catalog numberJ64425.MF
Price (USD)/ Each
448.00
-
Add to cart
Specifications
Chemical Name or MaterialLocostatin
CAS133812-16-5
ColorWhite
Recommended StorageKeep cold; Store under Argon
TSCANo
View more
Locostatin is a cell permeable, potent inhibitor of Raf kinase inhibitor protein (RKIP)/Raf1 kinase interaction and an inhibitor of cell migration

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Locostatin is a cell-permeable, potent inhibitor of Raf kinase inhibitor protein (RKIP)/Raf1 kinase interaction and an inhibitor of cell migration
RUO – Research Use Only