Thermo Scientific Chemicals

Carbetapentane citrate, Thermo Scientific Chemicals

Catalog number: J64446.MF
50 mg, Each
Thermo Scientific Chemicals

Carbetapentane citrate, Thermo Scientific Chemicals

Catalog number: J64446.MF
50 mg, Each
Quantity
Catalog number: J64446.MF
also known as J64446-MF
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
23142-01-0
IUPAC Name
2-[2-(diethylamino)ethoxy]ethyl 1-phenylcyclopentane-1-carboxylate; 2-hydroxypropane-1,2,3-tricarboxylic acid
Molecular Formula
C26H39NO10
InChI Key
AKJDEXBCRLOVTH-UHFFFAOYSA-N
SMILES
OC(=O)CC(O)(CC(O)=O)C(O)=O.CCN(CC)CCOCCOC(=O)C1(CCCC1)C1=CC=CC=C1
Assay (HPLC)
95.0% min.

Description

Sigma receptor agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text