Thermo Scientific Chemicals

5-Bromo-4-chloro-3-indolyl phosphate disodium salt, 98%, Thermo Scientific Chemicals

Catalog number: J64451.MD
250 mg, Each
Thermo Scientific Chemicals

5-Bromo-4-chloro-3-indolyl phosphate disodium salt, 98%, Thermo Scientific Chemicals

Catalog number: J64451.MD
250 mg, Each
Quantity
Catalog number: J64451.MD
also known as J64451-MD
Price (USD)
Quantity
-
Have Questions?

Description

5-Bromo-4-chloro-3-indolyl phosphate disodium salt, is used for the colorimetric detection of alkaline phosphatase-labeled molecules, 5-Bromo-4-chloro-3-indolyl phosphate (BCIP) and Nitro Blue Tetrazolium (NBT) are available. The BCIP/NBT substrate system is versatile and functions in a variety of applications, including Northern Southern, and Western blotting, in situ hybridization, and immunohistochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-4-chloro-3-indolyl phosphate disodium salt, is used for the colorimetric detection of alkaline phosphatase-labeled molecules, 5-Bromo-4-chloro-3-indolyl phosphate (BCIP) and Nitro Blue Tetrazolium (NBT) are available. The BCIP/NBT substrate system is versatile and functions in a variety of applications, including Northern Southern, and Western blotting, in situ hybridization, and immunohistochemistry.

Solubility
Soluble in water (50 mg/mL). Insoluble in dimethylformamide.

Notes
Moisture & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text