Sirtinol
Thermo Scientific Chemicals

Sirtinol

 
Have Questions?
Quantity:
10 mg
Catalog number J64486.MA
also known as J64486-MA
Price (USD)/ Each
209.00
-
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (USD)/ Each
209.00
Add to cart
Sirtinol
Catalog numberJ64486.MA
Price (USD)/ Each
209.00
-
Add to cart
Specifications
Quantity10 mg
DescriptionOff-white to Yellow
For Use With (Application)Sirtinol is a cell permeable specific inhibitor of sirtuin NAD-dependant deacetylases
Recommended StorageStore at -20°C
CAS410536-97-9
View more
Sirtinol is a cell permeable specific inhibitor of sirtuin NAD-dependant deacetylases

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C