PTP1B Inhibitor
Thermo Scientific Chemicals

PTP1B Inhibitor

 
Have Questions?
Quantity:
5 mg
Catalog number J64512.LB0
also known as J64512-#0
Price (USD)/ Each
494.00
-
Add to cart
Quantity:
5 mg
Request bulk or custom format
Price (USD)/ Each
494.00
Add to cart
PTP1B Inhibitor
Catalog numberJ64512.LB0
Price (USD)/ Each
494.00
-
Add to cart
Chemical Identifiers
CAS765317-72-4
IUPAC Name3-(3,5-dibromo-4-hydroxybenzoyl)-2-ethyl-N-{4-[(1,3-thiazol-2-yl)sulfamoyl]phenyl}-1-benzofuran-6-sulfonamide
Molecular FormulaC26H19Br2N3O7S3
InChI KeySXKBTDJJEQQEGE-UHFFFAOYSA-N
SMILESCCC1=C(C(=O)C2=CC(Br)=C(O)C(Br)=C2)C2=CC=C(C=C2O1)S(=O)(=O)NC1=CC=C(C=C1)S(=O)(=O)NC1=NC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)95.0% min.
PTP1B Inhibitor is used as a PTP1B inhibitor blocks the activity of the Protein Tyrosine Phosphatase 1B, which is a key factor in the negative regulation of insulin and leptin signaling pathway. Protein tyrosine phosphatase 1B (PTP1B) has critical roles in cell signaling relevant to diabetes, obesity, and cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • PTP1B inhibitor blocks the activity of the Protein Tyrosine Phosphatase 1B, which is a key factor in the negative regulation of insulin and leptin signaling pathway
RUO – Research Use Only
Contents & Storage
-30°C to -10°C