Azadibenzocyclooctyne acid, Thermo Scientific Chemicals
Azadibenzocyclooctyne acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Azadibenzocyclooctyne acid, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
Catalog number J64549.MB
also known as J64549-MB
Price (USD)/ Each
172.00
-
Add to cart
Quantity:
25 mg
Request bulk or custom format
Price (USD)/ Each
172.00
Add to cart
Azadibenzocyclooctyne acid, Thermo Scientific Chemicals
Catalog numberJ64549.MB
Price (USD)/ Each
172.00
-
Add to cart
Specifications
Chemical Name or MaterialAzadibenzocyclooctyne acid
CAS1425485-72-8
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Azadibenzocyclooctyne acid is the simplest amine reactive reagent for introducing a dibenzylcyclooctyne to available amine functionality.Azadibenzocyclooctyne acid is widely useful in strain-promoted copper-free azide-alkyne click chemistry reactions. It reacts with azide functionalized compounds or bimolecules to give stable triazole linkage without a need for a Cu(I) catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azadibenzocyclooctyne acid is the simplest amine reactive reagent for introducing a dibenzylcyclooctyne to available amine functionality.Azadibenzocyclooctyne acid is widely useful in strain-promoted copper-free azide-alkyne click chemistry reactions. It reacts with azide functionalized compounds or bimolecules to give stable triazole linkage without a need for a Cu(I) catalyst.

Solubility
Soluble in dimethyl sulfoxide and dimethyl formamide.

Notes
Store in cool place. Incompatible with amines.
RUO – Research Use Only

General References:

  1. Patterson, D. M.; Prescher, J. A. Orthogonal bioorthogonal chemistries. Curr. Opin. Chem. Biol. 2015, 28, 141-149.
  2. Jung, S.; Yi, H. Integrated fabrication-conjugation approaches for biomolecular assembly and protein sensing with hybrid microparticle platforms and biofabrication - A focused minireview. Korean J. Chem. Eng. 2015, 32 (9), 1713-1719.