Deoxy-bigCHAP is used for membrane solubilization. It is easily removed by dialysis and its absorption at UV region is very low. N,N-Bis[3-(D-gluconamido)propyl]deoxycholamide is used for membrane solubilization and proteomic analysis. N,N-Bis[3-(D-gluconamido)propyl]deoxycholamide (deoxy-BigCHAP) is a non-ionic detergent that solubilizes cell membrane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Deoxy-bigCHAP is used for membrane solubilization. It is easily removed by dialysis and its absorption at UV region is very low. N,N-Bis[3-(D-gluconamido)propyl]deoxycholamide is used for membrane solubilization and proteomic analysis. N,N-Bis[3-(D-gluconamido)propyl]deoxycholamide (deoxy-BigCHAP) is a non-ionic detergent that solubilizes cell membrane.
Solubility
Soluble in water at 50mg/ml. Also soluble in DMSO or methanol.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
- Takeshi Masuda.; Masaru Tomita.; Yasushi Ishihama. Phase Transfer Surfactant-Aided Trypsin Digestion for Membrane Proteome Analysis. J. Proteome Res. 2008, 7 (2), 731-740.
- Ayumi Ishibashi Dr.; Naotoshi Nakashima Prof. Dr. Individual Dissolution of Single-Walled Carbon Nanotubes in Aqueous Solutions of Steroid or Sugar Compounds and Their Raman and Near-IR Spectral Properties. Chemistry - A European Journal. 2006, 12 (29), 13524-13530.