1x1 image pixel for data collection
Deoxy-bigCHAP
Deoxy-bigCHAP
Deoxy-bigCHAP
Thermo Scientific Chemicals

Deoxy-bigCHAP

Analog of CHAPS and CHAPSO | CAS: 86303-23-3 | C42H75N3O15 | 862.068 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J64578.MD
also known as J64578-MD
Price (USD)
-
Quantity:
250 mg
Request bulk or custom format
Specifications
CAS86303-23-3
Chemical Name or MaterialDeoxy-bigCHAP
ColorWhite
Formula Weight862.05
Health Hazard 1H315-H319-H335
View more
Ask our AI about this Product
Deoxy-bigCHAP is used for membrane solubilization. It is easily removed by dialysis and its absorption at UV region is very low. N,N-Bis[3-(D-gluconamido)propyl]deoxycholamide is used for membrane solubilization and proteomic analysis. N,N-Bis[3-(D-gluconamido)propyl]deoxycholamide (deoxy-BigCHAP) is a non-ionic detergent that solubilizes cell membrane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Deoxy-bigCHAP is used for membrane solubilization. It is easily removed by dialysis and its absorption at UV region is very low. N,N-Bis[3-(D-gluconamido)propyl]deoxycholamide is used for membrane solubilization and proteomic analysis. N,N-Bis[3-(D-gluconamido)propyl]deoxycholamide (deoxy-BigCHAP) is a non-ionic detergent that solubilizes cell membrane.

Solubility
Soluble in water at 50mg/ml. Also soluble in DMSO or methanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Takeshi Masuda.; Masaru Tomita.; Yasushi Ishihama. Phase Transfer Surfactant-Aided Trypsin Digestion for Membrane Proteome Analysis. J. Proteome Res. 2008, 7 (2), 731-740.
  2. Ayumi Ishibashi Dr.; Naotoshi Nakashima Prof. Dr. Individual Dissolution of Single-Walled Carbon Nanotubes in Aqueous Solutions of Steroid or Sugar Compounds and Their Raman and Near-IR Spectral Properties. Chemistry - A European Journal. 2006, 12 (29), 13524-13530.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
P02E049Certificate of AnalysisDec 06, 2021J64578.MD, J64578.03
X11E014Certificate of AnalysisDec 06, 2021J64578.MD, J64578.03
Z12A038Certificate of AnalysisDec 06, 2021J64578.MD, J64578.03
3 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection