Thermo Scientific Chemicals

2'-Amino-2'-deoxyguanosine, 98%

Catalog number: J64650.03
1 g, Each
Thermo Scientific Chemicals

2'-Amino-2'-deoxyguanosine, 98%

Catalog number: J64650.03
1 g, Each
Quantity
Catalog number: J64650.03
also known as J64650-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
60966-26-9
IUPAC Name
2-amino-9-[(2R,3R,4S,5R)-3-amino-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
Molecular Formula
C10H14N6O4
InChI Key
ROPTVRLUGSPXNH-SJLGBTOWNA-N
SMILES
N[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C1N=C(N)NC2=O
Assay from Supplier's CofA
≥97.5% (HPLC)
Appearance (Color)
White to pale yellow to pale cream
Form
Powder

Description

2‘-Amino-2‘-deoxyguanosine is a valuable probe for investigating the metal ion interactions at the active site of the group I ribozyme.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2‘-Amino-2‘-deoxyguanosine is a valuable probe for investigating the metal ion interactions at the active site of the group I ribozyme.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text