Albumin, Bovine, Fraction V, 97%, Standard Grade, pH 7.0
This product is animal origin and can only be shipped within the United States
Albumin, Bovine, Fraction V, 97%, Standard Grade, pH 7.0
Thermo Scientific Chemicals

Albumin, Bovine, Fraction V, 97%, Standard Grade, pH 7.0

Albumin bovine/fraction V, CAS # 9048-46-8, also known as bovine serum albumin (BSA), is a serum albumin protein derived from cows.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1kg
10g
50g
100g
250g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J64655.A1
also known as J64655-A1
Price (USD)/ Each
1,158.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1363.00 
Save 204.35 (15%)
-
Add to cart
Quantity:
1kg
Request bulk or custom format
Price (USD)/ Each
1,158.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1363.00 
Save 204.35 (15%)
Add to cart
Albumin, Bovine, Fraction V, 97%, Standard Grade, pH 7.0
Catalog numberJ64655.A1
Price (USD)/ Each
1,158.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1363.00 
Save 204.35 (15%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
FormulationLyophilized powder
Recommended StorageKeep cold
pH Range7.0
EINECS Number232-936-2
Quantity1kg
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Albumin bovine/fraction V is a serum albumin protein commonly used in laboratory experiments including, enzyme-linked immunosorbent assay (ELISAs), immunoblot, and immune-histochemistry or -fluorescence

Applications

  • Albumin bovine/fraction V displays blocking activity mainly when casein-sensitive antibodies, including phospho-specific antibodies, are used
  • In genetic approaches this compound can be used to increase polymerase chain reactions (PCR) yields from low purity templates
  • This compound is useful for stabilizing enzymes during storage and also for enzymatic reactions where the absence of nucleases is crucial
  • In in vitro cultures, albumin bovine/fraction V has been applied as a nutrient in cell and microbial cultures especially when serum-free media is used
  • It is commonly used as a protein standard to obtain calibration curves for several laboratory experiments
  • Albumin bovine/fraction V is a small protein commonly used in immunohistochemistry to block nonspecific regions. The binding of this compound to nonspecific binding sites increases the probability of the antibodies bind only to the region of interest.
RUO – Research Use Only