Thermo Scientific Chemicals

1,4-Dithioerythritol, Electrophoresis Grade, 99%, Thermo Scientific Chemicals

Catalog number: J64656.03
1 g, Each
Thermo Scientific Chemicals

1,4-Dithioerythritol, Electrophoresis Grade, 99%, Thermo Scientific Chemicals

Catalog number: J64656.03
1 g, Each
Quantity
Catalog number: J64656.03
also known as J64656-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6892-68-8
IUPAC Name
(2R,3S)-1,4-disulfanylbutane-2,3-diol
Molecular Formula
C4H10O2S2
InChI Key
VHJLVAABSRFDPM-ZXZARUISSA-N
SMILES
O[C@@H](CS)[C@H](O)CS
Assay from Supplier's CofA
≥98.5% (titration)
Appearance (Color)
White
Form
Crystals or crystalline powder
UV/Vis
UV Ratios: A283 (0.2 M Aqueous solution): ≤0.04

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text