Thermo Scientific Chemicals

Choline bitartrate, 98+%, Thermo Scientific Chemicals

Catalog number: J64657.A1
1 kg, Each
Thermo Scientific Chemicals

Choline bitartrate, 98+%, Thermo Scientific Chemicals

Catalog number: J64657.A1
1 kg, Each
Quantity
Catalog number: J64657.A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
87-67-2
IUPAC Name
(2-hydroxyethyl)trimethylazanium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate
Molecular Formula
C9H19NO7
InChI Key
QWJSAWXRUVVRLH-LREBCSMRSA-M
SMILES
C[N+](C)(C)CCO.O[C@H]([C@@H](O)C([O-])=O)C(O)=O
Appearance (Color)
White
pH
3-4
Assay from Supplier's CofA
≥98.0%
Water Content
≤1.0%
Form
Powder or crystals or crystalline powder

Description

Choline Bitartrate is used by nerve cells to manufacture a chemical called acetylcholine, which effectively allows cells to communicate with each other. Choline is also heavily associated with neurological function and can help improve movement, coordination, and myofibril contraction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Choline Bitartrate is used by nerve cells to manufacture a chemical called acetylcholine, which effectively allows cells to communicate with each other. Choline is also heavily associated with neurological function and can help improve movement, coordination, and myofibril contraction.

Solubility
Soluble in alcohol and slightly soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text