Thermo Scientific Chemicals

Tropine DL-tropate, 99%, Thermo Scientific Chemicals

Catalog number: J64677.03
1 g, Each
Thermo Scientific Chemicals

Tropine DL-tropate, 99%, Thermo Scientific Chemicals

Catalog number: J64677.03
1 g, Each
Quantity
Catalog number: J64677.03
also known as J64677-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
51-55-8
IUPAC Name
8-methyl-8-azabicyclo[3.2.1]octan-3-yl 3-hydroxy-2-phenylpropanoate
Molecular Formula
C17H23NO3
InChI Key
RKUNBYITZUJHSG-UHFFFAOYNA-N
SMILES
CN1C2CCC1CC(C2)OC(=O)C(CO)C1=CC=CC=C1
Melting Point
114-118°C
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay from Supplier's CofA
≥98.5
Water Content (Karl Fischer Titration)
≤0.2%

Description

Tropine DL-tropate is used as a reduces brain levels of acetylcholine, Competitive nonselective antagonist at central and peripheral muscarinic acetylcholine receptors, used in opthalmology to dilate the pupil and in efforts of resuscitation in the treatment of bradycardia and cardiac arrest.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text