Thermo Scientific Chemicals
Cdc2-Like Kinase Inhibitor, TG003
Catalog number: J64690.LB0
5 mg, Each
Thermo Scientific Chemicals
Cdc2-Like Kinase Inhibitor, TG003
Catalog number: J64690.LB0
5 mg, Each
Quantity
Catalog number: J64690.LB0
also known as J64690-#0
Price (USD)
Quantity
-
Have Questions?
Description
Cdc2-Like Kinase Inhibitor, TG003 is a benzothiazole agent that acts as a specific and ATP-competitive inhibitor of Clk family members. Reports show that TG003 modifies the regulation of alternative splicing modulated by phosphorylation of SR proteins in vitro, and rescues defects in the development of Xenopus caused by an overabundance of Clk activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TG003 is a benzothiazole agent that acts as a specific and ATP-competitive inhibitor of Clk family members.