Thermo Scientific Chemicals

Glycerol, ultrapure, 99.5+%, Thermo Scientific Chemicals

Catalog number: J64719.AP
500 mL, Each
Thermo Scientific Chemicals

Glycerol, ultrapure, 99.5+%, Thermo Scientific Chemicals

Catalog number: J64719.AP
500 mL, Each
Quantity
Catalog number: J64719.AP
also known as J64719-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56-81-5
IUPAC Name
propane-1,2,3-triol
Molecular Formula
C3H8O3
InChI Key
PEDCQBHIVMGVHV-UHFFFAOYSA-N
SMILES
OCC(O)CO
Elemental Analysis
Chloride: ≤ 10 ppm
Assay (unspecified)
≥ 99.5%
Assay (unspecified)
Sulfates: ≤ 20 ppm
Heavy Metals
≤ 5 ppm as lead
Specific Gravity
≥ 1.249

Description

Glycerol, ultrapure is used in sample preparation and gel formation for polyacrylamide gel electrophoresis. Glycerol (5-10%) increases the density of a sample so that the sample will layer at the bottom of a gel's sample well. Glycerol is also used to aid in casting gradient gels and as a protein stabilizer and storage buffer component, glycerol is used as a solvent, plasticizer and humectant. Additional uses include as an aid in casting gradient gels, a protein stabilizer and storage buffer component.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text