Thermo Scientific Chemicals

Bismarck Brown R

Catalog number: J64757.14
25 g, Each
Thermo Scientific Chemicals

Bismarck Brown R

Catalog number: J64757.14
25 g, Each
Quantity
Catalog number: J64757.14
also known as J64757-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5421-66-9
IUPAC Name
4-(2-{3-[2-(2,4-diamino-5-methylphenyl)diazen-1-yl]-4-methylphenyl}diazen-1-yl)-6-methylbenzene-1,3-diamine dihydrochloride
Molecular Formula
C21H26Cl2N8
InChI Key
WLKAMFOFXYCYDK-UHFFFAOYSA-N
SMILES
Cl.Cl.CC1=CC(N=NC2=CC(N=NC3=C(N)C=C(N)C(C)=C3)=C(C)C=C2)=C(N)C=C1N
Appearance (Color)
Brownish-black
Loss on Drying
<1.0% w/w at 60?C
lambda-max (UV-Vis)
468nm in 50% ethanol
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text