Tripeptidyl Peptidase Inhibitor
Tripeptidyl Peptidase Inhibitor
Thermo Scientific Chemicals
Tripeptidyl Peptidase Inhibitor
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
10 mg
Catalog number J64760.LB0
also known as J64760-#0
Price (USD)/ Each
132.00
-
Add to cart
Quantity:
5 mg
Request bulk or custom format
Price (USD)/ Each
132.00
Add to cart
Tripeptidyl Peptidase Inhibitor
Catalog numberJ64760.LB0
Price (USD)/ Each
132.00
-
Add to cart
Specifications
Quantity5 mg
DescriptionOff-white
For Use With (Application)A serine protease inhibitor that acts as a potent, selective, dose-dependent, irreversible inhibitor of tripeptidylpeptidase II
Recommended StorageStore at -20°C; Store under Argon
CAS184901-82-4
View more
Tripeptidyl peptidase Inhibitor is a serine protease inhibitor which irreversibly inhibits TPPII (tripeptidyl peptidase II), a giant protease which may substitute for some proteasome functions, and reversibly inhibits Cln (TPPI). AAF-CMK shows only marginal inhibition of the chymotrypsin-like activity of the proteasome.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • A serine protease inhibitor that acts as a potent, selective, dose-dependent, irreversible inhibitor of tripeptidylpeptidase II
RUO – Research Use Only
Contents & Storage
-30°C to -10°C