Thermo Scientific Chemicals

Autophagy Inhibitor, 3-MA

Catalog number: J64813.MB
25 mg, Each
Thermo Scientific Chemicals

Autophagy Inhibitor, 3-MA

Catalog number: J64813.MB
25 mg, Each
Quantity
Catalog number: J64813.MB
also known as J64813-MB
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5142-23-4
IUPAC Name
3-methyl-3H-purin-6-amine
Molecular Formula
C6H7N5
InChI Key
FSASIHFSFGAIJM-UHFFFAOYSA-N
SMILES
CN1C=NC(N)=C2N=CN=C12
Form
Solid
Purity
≥99%
Appearance (Color)
White

Description

A synthetic intermediate and a cell-permeable autophagic sequestration blocker that protects cerebellar granule cells from apoptosis post serum/potassium deprivation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A synthetic intermediate and a cell-permeable autophagic sequestration blocker that protects cerebellar granule cells from apoptosis post serum/potassium deprivation
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text