Thermo Scientific Chemicals

2',3'-Dideoxyinosine, 98%

Catalog number: J64833.MD
250 mg, Each
Thermo Scientific Chemicals

2',3'-Dideoxyinosine, 98%

Catalog number: J64833.MD
250 mg, Each
Quantity
Catalog number: J64833.MD
also known as J64833-MD
Price (USD)
Quantity
-
Have Questions?

Description

2?,3?-Dideoxyinosine is a potent anti-retroviral agent. It is most effective in combination therapy for the treatment of HIV and related lymphoma.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′,3′-Dideoxyinosine is a potent anti-retroviral agent. It is most effective in combination therapy for the treatment of HIV and related lymphoma.

Solubility
Soluble in DMSO or methanol

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text