Thermo Scientific Chemicals

N-Succinimidyl 15-azido-4,7,10,13-tetraoxapentadecanoate, 90+%, Thermo Scientific Chemicals

Catalog number: J64834.MC
100 mg, Each
Thermo Scientific Chemicals

N-Succinimidyl 15-azido-4,7,10,13-tetraoxapentadecanoate, 90+%, Thermo Scientific Chemicals

Catalog number: J64834.MC
100 mg, Each
Quantity
Catalog number: J64834.MC
also known as J64834-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
944251-24-5
IUPAC Name
2,5-dioxopyrrolidin-1-yl 1-azido-3,6,9,12-tetraoxapentadecan-15-oate
Molecular Formula
C15H24N4O8
InChI Key
OIGKWPIMJCPGGD-UHFFFAOYSA-N
SMILES
[N-]=[N+]=NCCOCCOCCOCCOCCC(=O)ON1C(=O)CCC1=O
Appearance (Color)
Pale yellow to yellow
Thin Layer Chromatography
Single spot (DCM:MeOH 10:1)
Form
Liquid
Assay from Supplier's CofA
≥90.0%

Description

N-Succinimidyl 15-azido-4,7,10,13-tetraoxapentadecanoate is an amine reactive reagent for azide incorporation. It contains a PEG spacer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Succinimidyl 15-azido-4,7,10,13-tetraoxapentadecanoate is an amine reactive reagent for azide incorporation. It contains a PEG spacer.

Solubility
Soluble in dimethyl sulfoxide and acetonitrile.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text