Thermo Scientific Chemicals
L-Glutamic acid alpha-(7-amido-4-methylcoumarin), Thermo Scientific Chemicals
Catalog number: J64847.MF
50 mg, Each
Thermo Scientific Chemicals
L-Glutamic acid alpha-(7-amido-4-methylcoumarin), Thermo Scientific Chemicals
Catalog number: J64847.MF
50 mg, Each
Quantity
Catalog number: J64847.MF
also known as J64847-MF
Price (USD)
Quantity
-
Have Questions?
Description
A fluorogenic substrate for aminopeptidase a, producing a blue fluorescent solution upon cleavage.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
A fluorogenic substrate for aminopeptidase a, producing a blue fluorescent solution upon cleavage.