1,1'-Azobis(N,N-dimethylformamide), 95%, Thermo Scientific Chemicals
1,1'-Azobis(N,N-dimethylformamide), 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,1'-Azobis(N,N-dimethylformamide), 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number J64865.03
also known as J64865-03
Price (USD)/ Each
126.65
Online exclusive
140.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
126.65
Online exclusive
140.00 
Save 13.35 (10%)
Add to cart
1,1'-Azobis(N,N-dimethylformamide), 95%, Thermo Scientific Chemicals
Catalog numberJ64865.03
Price (USD)/ Each
126.65
Online exclusive
140.00 
Save 13.35 (10%)
-
Add to cart
Specifications
Chemical Name or Material1,1'-Azobis(N,N-dimethylformamide)
CAS10465-78-8
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Melting Point112°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only